missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPAG17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555938
This item is not returnable.
View return policy
Description
Immunogen sequence: DLTPLLSFQA TDPVNGTVMT TREDKVVIVE RKDGTRIVDH ADGTRITTFY QVYEDQIILP DDQETTEGPR TVTRQVKCMR VESSRYATVI ANCEDSSCCA TF Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
Specifications
| SPAG17 | |
| Polyclonal | |
| Unconjugated | |
| SPAG17 | |
| 4931427F14Rik; CT143; PF6; projection protein PF6 homolog; SPAG17; Spag17-ps; sperm associated antigen 17; sperm associated antigen 17, pseudogene; sperm-associated antigen 17 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 200162 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6Q759 | |
| SPAG17 | |
| Recombinant protein corresponding to Human SPAG17. Recombinant protein control fragment (Product #RP-94344). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction