missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPAG4L Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05670-100ul
This item is not returnable.
View return policy
Description
SPAG4L Polyclonal antibody specifically detects SPAG4L in Human, Mouse samples. It is validated for Western Blot
Specifications
| SPAG4L | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| dJ726C3.1, MGC33594, Sad1 and UNC84 domain containing 5, SPAG4L, Sperm-associated antigen 4-like protein, SUN domain-containing protein 5, testis and spermatogenesis related gene 4, Testis and spermatogenesis-related gene 4 protein, TSARG4sperm associated antigen 4-like | |
| Recombinant protein of mouse SPAG4L | |
| 100 μg | |
| Cell Biology | |
| 140732 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction