missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPAG5 Monoclonal Antibody (OTI2D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810453
This item is not returnable.
View return policy
Description
A novel gene product, hMAP126, interacts with p29 in the yeast two-hybrid assay. The subcellular distribution of hMAP126 was localized to the mitotic spindle and is phosphorylated by p34(cdc2) kinase. Human MAP126 is likely involved in the functional and dynamic regulation of mitotic spindles.Specifications
| SPAG5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96R06 | |
| SPAG5 | |
| Human recombit protein fragment corresponding to amino acids 1-297 of human SPAG5 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2D6 | |
| Unconjugated | |
| SPAG5 | |
| DEEPEST, MAP126, hMAP126 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10615 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction