missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPAG5 Monoclonal Antibody (OTI3F9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810450S
This item is not returnable.
View return policy
Description
SPAG5 Monoclonal antibody specifically detects SPAG5 in Human samples. It is validated for Western BlotSpecifications
| SPAG5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96R06 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10615 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI3F9 | |
| Unconjugated | |
| SPAG5 | |
| AI874642; Astrin; D11Bhm180e; Deepest; hMAP126; MAP126; Mastrin; microtubule-associated protein; mitotic spindle associated protein p126; mitotic spindle coiled-coil related protein; mitotic spindle-associated protein p126; S17; Spag5; sperm associated antigen 5; sperm-associated antigen 5 | |
| Human recombinant protein fragment corresponding to amino acids 1-297 of human SPAG5 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction