missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPAG7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570770
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 92%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
SPAG7 gene ontology annotations related to this gene include nucleic acid binding.
Specifications
| SPAG7 | |
| Polyclonal | |
| Unconjugated | |
| SPAG7 | |
| 5730443G10; ACRP; FSA-1; Fsa1l; hypothetical protein LOC515086; similar to sperm associated antigen 7; SPAG7; sperm associated antigen 7; sperm-associated antigen 7; Sperm-associated antigen 7 homolog; zgc:66397 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 216873 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q7TNE3 | |
| SPAG7 | |
| synthetic peptide directed towards the N-terminal region of mouse SPAG7. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction