missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SPAG8 Polyclonal antibody specifically detects SPAG8 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | SPAG8 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | BS-84, HSD-1MGC26201, hSMP-1, SMP1, SMP-1, SPAG3, sperm associated antigen 8, Sperm membrane protein 1, Sperm membrane protein BS-84, sperm-associated antigen 8 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 220-420 of human SPAG8 (NP_758516.1).,, Sequence:, LVADRVPNYTSWSQHCPWEPQKQPPWEFLQVLEPGARGLWKPPDIKGKLMVCYETLPRGQCLLYNWEEERATNHLDQVPSMQDGSESFFFRHGHRGLLTMQLKSPMPSSTTQKDSYQPPGNVYWPLRGKREAMLEMLLQHQICKEVQAEQEPTRKLFEVESVTHHDYRMELAQAGTPAPTKPHDYRQEQPETFWIQRAPQL |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?