missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SPAG9 Antibody [Allophycocyanin], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB100-79793APC
This item is not returnable.
View return policy
Description
SPAG9 Polyclonal antibody specifically detects SPAG9 in Human,Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| SPAG9 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| APC | |
| Cancer/testis antigen 89, c-Jun NH2-terminal kinase-associated leucine zipper protein, C-Jun-amino-terminal kinase-interacting protein 4, CT89JIP4, FLJ14006, FLJ26141, FLJ34602, HLC4, HLC-6, HSS, Human lung cancer oncogene 6 protein, JLPPHETSYD1FLJ13450, JNK interacting protein, JNK/SAPK-associated protein, JNK-associated leucine-zipper protein, JNK-interacting protein 4, KIAA0516JIP-4, lung cancer oncogene 4, MAPK8IP4, Max-binding protein, MGC117291, MGC14967, MGC74461, Mitogen-activated protein kinase 8-interacting protein 4, PIG6, proliferation-inducing gene 6, Proliferation-inducing protein 6, Protein highly expressed in testis, sperm associated antigen 9, Sperm surface protein, Sperm-associated antigen 9, Sperm-specific protein, Sunday driver 1 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human JNK/SAPK-Associated Protien using the numbering given in entry NP_003962.3 (GeneID 9043) | |
| 0.1 mL | |
| Cell Cycle and Replication, Signal Transduction | |
| 9043 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction