missing translation for 'onlineSavingsMsg'
Learn More

SPANXN3 Antibody (OTI4B1), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18181006 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18181006 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18181006 Supplier Novus Biologicals Supplier No. NBP274308AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SPANXN3 Monoclonal specifically detects SPANXN3 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPANXN3
Applications Western Blot
Classification Monoclonal
Clone OTI4B1
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cancer/testis antigen family 11, member 8, CT11.8, Nuclear-associated protein SPAN-Xn3, SPANX family member N3, SPANX family, member N3, SPANX-N3sperm protein associated with the nucleus on the X chromosome N3
Gene Symbols SPANXN3
Host Species Mouse
Immunogen Full length recombinant protein of human SPANXN3 (NP_001009609) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 139067.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.