missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPANXN3 Antibody (OTI4B1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74308
This item is not returnable.
View return policy
Description
SPANXN3 Monoclonal specifically detects SPANXN3 in Human samples. It is validated for Western Blot.
Specifications
| SPANXN3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500 | |
| cancer/testis antigen family 11, member 8, CT11.8, Nuclear-associated protein SPAN-Xn3, SPANX family member N3, SPANX family, member N3, SPANX-N3sperm protein associated with the nucleus on the X chromosome N3 | |
| Mouse | |
| 15.4 kDa | |
| 100 μg | |
| 139067 | |
| Human | |
| Lyophilized |
| Western Blot | |
| OTI4B1 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| SPANXN3 | |
| Full length human recombinant protein of human SPANXN3 (NP_001009609) produced in E.coli. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction