missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPANXN3 Monoclonal Antibody (1F11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524522
This item is not returnable.
View return policy
Description
Peptide Sequence: MEQPTSSTNG EKTKSPCESN NKKNDEMQEV PNRVLAPEQS LKKTKTSEYP IIFVYYLRKG KKINSNQLEN EQSQENSINP IQKEEDEGVD LSEGSSNEDE DLGPCEGPSK EDKDLDSSEG SSQEDEDLGL SEGSSQDSGE D.
SPANXN3 is a protein coding gene.
Specifications
| SPANXN3 | |
| Monoclonal | |
| Unconjugated | |
| SPANXN3 | |
| cancer/testis antigen family 11, member 8; CT11.8; Nuclear-associated protein SPAN-Xn3; SPANX family member N3; SPANX family, member N3; SPANXN3; SPANX-N3; Sperm protein associated with the nucleus on the X chromosome N3 | |
| Mouse | |
| Protein A | |
| RUO | |
| 139067 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1F11 | |
| PBS with no preservative; pH 7.4 | |
| Q5MJ09 | |
| SPANXN3 | |
| SPANXN3 (NP_001009609.1, 1 a.a. ~ 141 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction