missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPANXN3 Monoclonal Antibody (OTI4B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809059S
This item is not returnable.
View return policy
Description
SPANXN3 Monoclonal antibody specifically detects SPANXN3 in Human samples. It is validated for Western BlotSpecifications
| SPANXN3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75362 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7764 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4B1 | |
| Unconjugated | |
| ZNF217 | |
| cancer/testis antigen family 11, member 8; CT11.8; Nuclear-associated protein SPAN-Xn3; SPANX family member N3; SPANX family, member N3; SPANXN3; SPANX-N3; Sperm protein associated with the nucleus on the X chromosome N3 | |
| Human recombinant protein fragment corresponding to amino acids 730-1048 of human ZNF217 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction