Learn More
SPARC Antibody (122511), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB941R
Product Code. 30059364
  • € 652.00 / 0.10mL
Description
SPARC Monoclonal antibody specifically detects SPARC in Human samples. It is validated for ELISA, Immunohistochemistry, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SPARC
Monoclonal
Alexa Fluor 647
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry, Flow Cytometry, CyTOF
122511
ELISA, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin)
Mouse myeloma cell line NS0-derived recombinant human SPARC, Ala18-Ile303, Accession # P09486
0.1 mL
Biologically Active Proteins, Cancer, Cell Cycle and Replication
6678
Store at 4C in the dark.