missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPARC Antibody (122511) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB941L
This item is not returnable.
View return policy
Description
SPARC Monoclonal antibody specifically detects SPARC in Human samples. It is validated for ELISA, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| SPARC | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
| 122511 | |
| ELISA, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin) | |
| Mouse myeloma cell line NS0-derived recombinant human SPARC, Ala18-Ile303, Accession # P09486 | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, Cell Cycle and Replication | |
| 6678 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction