missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPARC Antibody (122511) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB941JF669
This item is not returnable.
View return policy
Description
SPARC Monoclonal antibody specifically detects SPARC in Human samples. It is validated for ELISA,Immunohistochemistry,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| SPARC | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin) | |
| Mouse myeloma cell line NS0-derived recombinant human SPARC, Ala18-Ile303, Accession # P09486 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
| 122511 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, Cell Cycle and Replication | |
| 6678 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction