missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPARC Antibody (124413), Janelia Fluor™ 549, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB942I
This item is not returnable.
View return policy
Description
SPARC Monoclonal antibody specifically detects SPARC in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Specifications
| SPARC | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin) | |
| Mouse myeloma cell line NS0-derived recombinant mouse SPARC/Osteonectin, Ala18-Ile302, Accession # P07214 | |
| 0.5 mL | |
| Biologically Active Proteins, Cancer, Cell Cycle and Replication | |
| 6678 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| 124413 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction