missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPARC Antibody (MM0557-8N38), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11913V
This item is not returnable.
View return policy
Description
SPARC Monoclonal specifically detects SPARC in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SPARC | |
| Monoclonal | |
| DyLight 405 | |
| P09486 | |
| SPARC | |
| Human recombinant SPARC | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 6678 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0557-8N38 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction