missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPARC Antibody (OSTN/8528R), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20949AF350
This item is not returnable.
View return policy
Description
SPARC Monoclonal antibody specifically detects SPARC in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SPARC | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin) | |
| Recombinant fragment (around aa1-200) of human SPARC protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, Cell Cycle and Replication | |
| 6678 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| OSTN/8528R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction