missing translation for 'onlineSavingsMsg'
Learn More

SPARC Antibody (rOSTN/8527), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-20948G

Product Code. 18583269

  • € 728.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SPARC Monoclonal antibody specifically detects SPARC in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SPARC
Monoclonal
DyLight 488
Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin)
Recombinant fragment (around aa1-200) of human SPARC protein (exact sequence is proprietary)
0.1 mL
Biologically Active Proteins, Cancer, Cell Cycle and Replication
6678
Store at 4°C in the dark.
IgG1 κ
Immunohistochemistry (Paraffin)
rOSTN/8527
50 mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.