Learn More
SPARC Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15274-1-AP-20UL
 View more versions of this product
Product Code. 16894374
  • € 145.00 / 20µL
Description
SPARC is a secreted protein, acidic and rich in cysteines. It is a matrix-associated protein that elicits changes in cell shape, inhibits cell-cycle progression, and influences the synthesis of extracellular matrix (ECM)
Specifications
Specifications
SPARC
Polyclonal
Unconjugated
SPARC
Basement membrane protein 40, BM 40, ON, Osteonectin, SPARC
Rabbit
Antigen Affinity Chromatography
RUO
20692, 24791, 595124, 6678
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07214, P09486, P16975, P20112
SPARC
SPARC Fusion Protein Ag7390
20 μL
Primary
Human, Mouse, Rat, Pig