Learn More
Hirschmann™ Spare Burettes
Spare part for the economical repair of a Schilling burette.
Brand:  Hirschmann™ 8900775
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10576722
  • € 53.70 / Each
Estimated Shipment: 17-06-2024
to see stock.
Add to basket
Description
Description
  • For Schilling burettes
Specifications
Specifications
50 mL
Schilling Titration Units
1 Each
330 mm
0.1 mL
Automatic Buret