missing translation for 'onlineSavingsMsg'
Learn More

Poulten & Graf GmbH Spare Piston for FORTUNA™ OPTIFIX™ BASIC Dispenser

Product Code. 10289761
Change view
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
30 mL
50 mL
100 mL
200 mL
300 mL
500 mL
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10289761 200 mL Each
10117531 300 mL Each
10197571 10 mL Each
10249861 100 mL Each
10416121 5 mL Each
10428611 500 mL Each
10535352 30 mL Each
10678592 50 mL Each
10781883 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 10289761 Supplier Poulten & Graf GmbH Supplier No. 101 00053

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Poulten & Graf GmbH Spare Piston for FORTUNA™ OPTIFIX™ BASIC Dispenser comes with a finger guard and volume setting system.

  • Solid PTFE encased
  • With finger guard and volume setting system

Specifications

Capacity (Metric) Media 200 mL
Description Replacement Piston for OPTIFIX BASIC Dispenser
Capacity (Metric) 200 mL
For Use With (Equipment) OPTIFIX BASIC Dispenser
No. per Case 1
No. per Pack 1
Product Type Spare parts for Dispensers
Autoclavable Autoclavable
Disposable Reusable
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.