Learn More
Poulten & Graf GmbH Spare Piston for FORTUNA™ OPTIFIX™ SAFETY Dispenser
Product Code. 10076091
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
30 mL
50 mL
100 mL
Unit Size:
Each
Product Code. 10076091
Brand: Poulten & Graf GmbH 101 00351
Only null left
Add to basket
Add to basket
Poulten & Graf GmbH Spare Piston for FORTUNA™ OPTIFIX™ SAFETY Dispenser comes with a finger guard and volume setting system.
  • Solid PTFE encased
  • With finger guard and volume setting system
  • With sealing lip
Specifications
Capacity (Metric) Media 100 mL
Description Piston for OPTIFIX SAFETY
Capacity (Metric) 100 mL
For Use With (Equipment) OPTIFIX SAFETY Bottle top dispenser
No. per Case 1
No. per Pack 1
Product Type Spare part for bottle top dispenser
Autoclavable Not Autoclavable
Disposable Reusable