Learn More
Poulten & Graf GmbH Spare Piston for FORTUNA™ OPTIFIX™ SAFETY Dispenser
Product Code. 10066091
Change view
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
30 mL
50 mL
100 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10066091 50 mL Each
10076091 100 mL Each
10086091 10 mL Each
10229621 5 mL Each
10239911 30 mL Each
10371122 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10066091 Supplier Poulten & Graf GmbH Supplier No. 101 00348
Poulten & Graf GmbH Spare Piston for FORTUNA™ OPTIFIX™ SAFETY Dispenser comes with a finger guard and volume setting system.
  • Solid PTFE encased
  • With finger guard and volume setting system
  • With sealing lip
Specifications
Capacity (Metric) Media 50 mL
Description Piston for OPTIFIX SAFETY
Capacity (Metric) 50 mL
For Use With (Equipment) OPTIFIX SAFETY Bottle top dispenser
No. per Case 1
No. per Pack 1
Product Type Spare part for bottle top dispenser
Autoclavable Not Autoclavable
Disposable Reusable