missing translation for 'onlineSavingsMsg'
Learn More

Poulten & Graf GmbH Spare Piston for FORTUNA™ OPTIFIX™ SAFETY Dispenser

Product Code. 10371122
Change view
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
30 mL
50 mL
100 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10371122 2 mL Each
10066091 50 mL Each
10076091 100 mL Each
10086091 10 mL Each
10229621 5 mL Each
10239911 30 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10371122 Supplier Poulten & Graf GmbH Supplier No. 101 00327

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Poulten & Graf GmbH Spare Piston for FORTUNA™ OPTIFIX™ SAFETY Dispenser comes with a finger guard and volume setting system.

  • Solid PTFE encased
  • With finger guard and volume setting system
  • With sealing lip

Specifications

Capacity (Metric) Media 2 mL
Description Piston for OPTIFIX SAFETY
Capacity (Metric) 2 mL
For Use With (Equipment) OPTIFIX SAFETY Bottle top dispenser
No. per Case 1
No. per Pack 1
Product Type Spare part for bottle top dispenser
Autoclavable Not Autoclavable
Disposable Reusable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.