Learn More
Poulten & Graf GmbH Spare Piston for FORTUNA™ OPTIFIX™ SAFETY Dispenser
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
30 mL
50 mL
100 mL
Unit Size:
Each
Description
- Solid PTFE encased
- With finger guard and volume setting system
- With sealing lip
Specifications
Specifications
| Capacity (Metric) Media | 50 mL |
| Description | Piston for OPTIFIX SAFETY |
| Capacity (Metric) | 50 mL |
| For Use With (Equipment) | OPTIFIX SAFETY Bottle top dispenser |
| No. per Case | 1 |
| No. per Pack | 1 |
| Product Type | Spare part for bottle top dispenser |
| Autoclavable | Not Autoclavable |
| Disposable | Reusable |