missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten & Graf GmbH Spare Piston for FORTUNA™ POLYFIX™ Dispenser
Click to view available options
Volume (Metric):
10 mL
50 mL
Description:
Spare Piston
Spare Piston, PTFE-coated
Unit Size:
Each
Description
- With finger guard and volume setting system
Specifications
Specifications
| Product Type | Spare Piston for POLYFIX Dispenser |
| Autoclavable | Autoclavable |
| Description | Spare Piston |
| Volume (Metric) | 50 mL |
| Quantity | 1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur