Learn More
Heidolph™ Spare Tension Roller
Product Code. 10351732 Shop All Heidolph Instruments Products
Change view
Click to view available options
For Use With (Equipment):
Heidolph Instruments
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10351732 Heidolph Instruments Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10351732 Supplier Heidolph™ Supplier No. 5497100000
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Unimax 1010, Duomax 1030, Polymax 1040 and Promax 1020
1 tension roller as accessory for attachment
Order Info
dimType
TRUSTED_SUSTAINABILITY
Specifications
Voltage 115/230 V
Product Type Tension Roller
For Use With (Equipment) Heidolph Instruments