Learn More
Poulten & Graf GmbH Spare Valve Block for FORTUNA™ OPTIFIX™
Product Code. p-7116446
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
100 mL
300 mL
missing translation for 'unitSize'
Each
Product Code. 10068232
missing translation for 'mfr': Poulten & Graf GmbH 101 00451
Poulten & Graf GmbH Spare Valve Block for FORTUNA™ OPTIFIX™ comes with dispensing cylinder and discharge tube.
  • For OPTIFIX Basic, Solvent, and Safety
  • Comes with dispensing cylinder
  • Includes metal spring
Specifications
Product Type Spare Valve Block for OPTIFIX Dispensers
Autoclavable Autoclavable
Description Spare Valve block for OPTIFIX dispensers
Volume (Metric) 100 mL
For Use With (Equipment) OPTIFIX dispensers
Quantity 1