Learn More
Poulten & Graf GmbH Spare Valve Block for FORTUNA™ OPTIFIX™
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
100 mL
300 mL
missing translation for 'unitSize'
Each
Description
- For OPTIFIX Basic, Solvent, and Safety
- Comes with dispensing cylinder
- Includes metal spring
Specifications
Specifications
| Product Type | Spare Valve Block for OPTIFIX Dispensers |
| Autoclavable | Autoclavable |
| Description | Spare Valve block for OPTIFIX dispensers |
| Volume (Metric) | 100 mL |
| For Use With (Equipment) | OPTIFIX dispensers |
| Quantity | 1 |