Learn More
Poulten & Graf GmbH Spare Valve Block Unit with Amber Dispensing Cylinder
Poulten & Graf GmbH Spare Valve Block Unit with Amber Dispensing Cylinder is designed to be used with FORTUNA™ POLYFIX™ and comes with discharge tubes.
Brand: Poulten & Graf GmbH 101 07948
Additional Details : Weight : 0.30000kg
Description
- Complete with amber dispensing cylinder
- Includes discharge tube
Specifications
Spare Parts for POLYFIX Dispenser | |
Spare valve block unit | |
1 |
Autoclavable | |
10 to 50 mL | |
GL 32 |