Learn More
Poulten & Graf GmbH Spare Valve Block Unit with Amber Dispensing Cylinder
Poulten & Graf GmbH Spare Valve Block Unit with Amber Dispensing Cylinder is designed to be used with FORTUNA™ POLYFIX™ and comes with discharge tubes.
Brand:  Poulten & Graf GmbH 101 07938
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 12683415
  • € 154.00 / Each
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
Description
Description
  • Complete with amber dispensing cylinder
  • Includes discharge tube
Specifications
Specifications
Spare Parts for POLYFIX Dispenser
Spare valve block unit
1
Autoclavable
2 to 10 mL
GL 32
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers