missing translation for 'onlineSavingsMsg'
Learn More

Sparfloxacin, MedChemExpress

Product Code. 16442096
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16442096 500 mg 500mg
16478226 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16442096 Supplier Medchem Express Supplier No. HYB0308

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sparfloxacin (CI-978) is a fluoroquinolone antibiotic, shows broad and potent antibacterial activity.

Sparfloxacin (CI-978) is a fluoroquinolone antibiotic, shows broad and potent antibacterial activity.

Specifications

Chemical Name or Material Sparfloxacin
Physical Form Powder
CAS 110871-86-8
Color Yellow
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C19H22F2N4O3
For Use With (Application) COVID-19-immunoregulation
Synonym CI-978 AT-4140
Solubility Information 0.1 M NaOH : 50 mg/mL (127.42 mM; ultrasonic and adjust pH to 11 with NaOH) ∣DMSO : 3.33 mg/mL (8.49 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CN(C2CC2)C3=C(C(N)=C(F)C(N4C[C@H](C)N[C@H](C)C4)=C3F)C1=O)O
Molecular Weight (g/mol) 392.4
Formula Weight 392.4
Percent Purity 98.07%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.