missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spastin Antibody (Sp 6C6), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50303AF700
This item is not returnable.
View return policy
Description
Spastin Monoclonal specifically detects Spastin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Spastin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SPAST | |
| Recombinant Spastin | |
| 0.1 mL | |
| Cardiovascular Biology, Cell Cycle and Replication, Cellular Signaling, Epigenetics, metabolism, Neuroscience | |
| 6683 | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Sp 6C6 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| ADPSPSpastic paraplegia 4 protein, FSP2EC 3.6.4.3, KIAA1083EC 3.6.1.3, spastic paraplegia 4 (autosomal dominant; spastin), spastin, SPG4 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu