missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spastin Antibody (Sp 6C6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 195.00 - € 361.00
Specifications
| Antigen | Spastin |
|---|---|
| Clone | Sp 6C6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500, ELISA 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18606446
|
Novus Biologicals
NBP2-50303-0.025ml |
0.025 mL |
€ 195.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600837
|
Novus Biologicals
NBP2-50303 |
0.1 mL |
€ 361.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Spastin Monoclonal antibody specifically detects Spastin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Spastin | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cardiovascular Biology, Cell Cycle and Replication, Cellular Signaling, Epigenetics, metabolism, Neuroscience | |
| PBS | |
| 6683 | |
| IgG2a | |
| Protein A purified |
| Sp 6C6 | |
| Western Blot 1:500, ELISA 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ADPSPSpastic paraplegia 4 protein, FSP2EC 3.6.4.3, KIAA1083EC 3.6.1.3, spastic paraplegia 4 (autosomal dominant; spastin), spastin, SPG4 | |
| Recombinant Spastin | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title