missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPATA12 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15858143
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15858143 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15858143 Supplier Invitrogen™ Supplier No. PA569734

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

This gene is expressed primarily in testis and may play a role in testicular development and spermatogenesis. The encoded protein may be upregulated in response to ultraviolet-C radiation. [provided by RefSeq, Dec 2015].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SPATA12
Gene Accession No. Q7Z6I5
Gene Alias SPATA12; spermatogenesis associated 12; spermatogenesis-associated protein 12; Spermatogenesis-related protein 5; SRG5; testicular tissue protein Li 184
Gene Symbols SPATA12
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human SPATA12.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353324
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.