missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA19 Antibody, DyLight 594, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-41149DL594
This item is not returnable.
View return policy
Description
SPATA19 Polyclonal antibody specifically detects SPATA19 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| SPATA19 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| 219938 | |
| Store at 4°C in the dark. | |
| IgY |
| Western Blot, ELISA | |
| DyLight 594 | |
| cancer/testis antigen 132, CT132, FLJ25851, SPAS1, spergen 1, spergen1, spergen-1, spermatogenesis associated 19, spermatogenesis-associated protein 19, mitochondrial, Spermatogenic cell-specific gene 1 protein, spermatogenic specific-gene1 | |
| Antibody was raised against an 17 amino acid synthetic peptide near the amino terminus of human SPATA19. The immunogen is located within amino acids 30 - 80 of SPATA19. Amino Acid Squence: CLKKTEEEASRGIKEKLS | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction