missing translation for 'onlineSavingsMsg'
Learn More

SPATA19 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15991395
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15991395 100 μg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15991395 Supplier Invitrogen Supplier No. PA572740

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SPATA19 Polyclonal Antibody

SPATA19 Polyclonal Antibody for Western Blot, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA19
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SPATA19
Gene Accession No. Q7Z5L4, Q920Q3, Q9DAQ9
Gene Alias 1700001L23Rik; AV047838; cancer/testis antigen 132; CT132; Spas1; SPATA19; spergen1; Spergen-1; spermatogenesis associated 19; spermatogenesis-associated protein 19, mitochondrial; Spermatogenic cell-specific gene 1 protein; spermatogenic specific-gene1; testis secretory sperm-binding protein Li 243mP
Gene Symbols SPATA19
Host Species Chicken
Immunogen SPATA19 antibody for an 17 amino acid synthetic peptide near the amino terminus of human SPATA19. The immunogen is within amino acids 30 - 80 of SPATA19.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171403, 219938, 75469
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.