missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA2 Antibody (Z1P1F10*F12), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50304MFV500
This item is not returnable.
View return policy
Description
SPATA2 Monoclonal antibody specifically detects SPATA2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| SPATA2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| Z1P1F10*F12 | |
| Western Blot, ELISA, Immunohistochemistry | |
| KIAA0757FLJ13167, PD1spermatogenesis associated PD1, spermatogenesis associated 2, spermatogenesis-associated protein 2, Spermatogenesis-associated protein PD1, tamo | |
| Peptide Sequence - KSTQLSHLVYR | |
| 0.1 mL | |
| Cancer, Cellular Signaling, Cytokines and Growth Factors | |
| 9825 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction