missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA25 Antibody (Z37P3H11*B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50305
This item is not returnable.
View return policy
Description
SPATA25 Monoclonal antibody specifically detects SPATA25 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| SPATA25 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| C20orf165, Chromosome 20 Open Reading Frame 165, DJ337O18.8, Spermatogenesis Associated 25, Testis-Specific Gene 23 Protein, TSG23 | |
| Peptide Sequence - LVRSKRGQP | |
| 0.1 mL | |
| Developmental Biology | |
| 128497 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| Z37P3H11*B2 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction