missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA2L Monoclonal Antibody (OTI2E2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802375S
This item is not returnable.
View return policy
Description
SPATA2L Monoclonal antibody specifically detects SPATA2L in Human samples. It is validated for Western BlotSpecifications
| SPATA2L | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8IUW3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 124044 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2E2 | |
| Unconjugated | |
| SPATA2L | |
| C16orf76; SPATA2L; SPATA2-like protein; spermatogenesis associated 2 like; spermatogenesis associated 2-like; spermatogenesis-associated protein 2-like protein; tamo | |
| Human recombinant protein fragment corresponding to amino acids 179-424 of human SPATA2L produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction