missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPATA31D1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561618
This item is not returnable.
View return policy
Description
Immunogen sequence: IRAAFTGTTE AQKIRKDTRE FLEEKLGHRH GIDITCPQEP LSFPVGLGKA QHNPEVHVRA EPVQGCPCNY RAPSCKVTRT KSCSQQAIFV GQNYPTRIRQ IIDKDRQPQK VEAFKGKILC QSHPQSMPHR KPVPHPNPTC R Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 26%.
Specifications
| SPATA31D1 | |
| Polyclonal | |
| Unconjugated | |
| SPATA31D1 | |
| FAM75D1; FAM75-like protein FLJ46321; family with sequence similarity 75, member D1; protein FAM75D1; SPATA31 subfamily D member 1; SPATA31 subfamily D, member 1; SPATA31D1; Spermatogenesis-associated protein 31D1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 389763 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZQQ2 | |
| SPATA31D1 | |
| Recombinant protein corresponding to Human SPATA31D1. Recombinant protein control fragment (Product #RP-90571). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction