missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA7 Polyclonal Antibody

SPATA7 Polyclonal Antibody
Brand: Invitrogen PA5100336
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
SPATA7 Polyclonal Antibody for Western Blot
Tekniske data
| SPATA7 | |
| Polyclonal | |
| Unconjugated | |
| SPATA7 | |
| AI661438; B230306G18Rik; epididymis secretory protein Li 296; fertility related protein WMP1; fertility-related protein WMP1; HEL-S-296; HSD3; HSD-3.1; LCA3; RSD-3; si:ch211-51o12.1; SPATA7; sperm DNA no.3; spermatogenesis associated 7; spermatogenesis-associated protein 7; Spermatogenesis-associated protein 7 homolog; spermatogenesis-associated protein 7 homolog; spermatogenesis-associated protein 7; spermatogenesis-associated protein HSD3; Wmp1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 104871, 55812 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q80VP2, Q9P0W8 | |
| SPATA7 | |
| A synthesized peptide derived from human SPATA7(Accession Q9P0W8), corresponding to amino acid residues A9-I59. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion