missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATC1L Antibody (OTI1H6), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72277JF646
This item is not returnable.
View return policy
Description
SPATC1L Monoclonal specifically detects SPATC1L in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| SPATC1L | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SPATC1L | |
| Full length recombinant protein of human C21orf56 (NP_115637) produced in HEK293T cells. | |
| 0.1 mL | |
| 84221.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI1H6 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| chromosome 21 open reading frame 56, DKFZp434N0650, MGC99490 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction