Description
SA-AFO is a combination of a 2-ply, white, heavy weight, bonded and perforated oil only sorbent roll (76 cm x 46 m, perfed every 76 cm) that absorbs oil and petroleum based liquids and a nitrile rubber anti-fatigue base mat (82 x 152 cm).
- Special design provides buoyancy for basic fatigue relief.
- Silicone free allowing mat to be used in vehicle painting facilities.
- High quality nitrile rubber makes the anti-fatique mat oil resistant.
- Designed to keeps workers safe while protecting the floor from oils and other industrial liquids.
Specifications
Specifications
| Color | Gray |
| Capacity (English) | 48.872 gal. |
| Capacity (Metric) | 185 L |
| Type | Absorbent Kit |
| For Use With (Application) | Beverage Processing, Chemical, Food Processing, Laboratory, Manufacturing, Maritime, Oil and Gas, Process, Warehouse and Logistics |
| Includes | One 76 cm x 46 m Perforated Roll and One Antifatigue Base Mat 82 x 152 cm |
| Model | Oil Only Sorbent |
| Alternate Part Number | SA-AFO |