missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPC18 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94187-0.02ml
This item is not returnable.
View return policy
Description
SPC18 Polyclonal antibody specifically detects SPC18 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| SPC18 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| EC 3.4, Endopeptidase SP18, Microsomal signal peptidase 18 kDa subunit, SEC11 homolog A, SEC11 homolog A (S. cerevisiae), SEC11L1, SEC11-like 1, SEC11-like protein 1, sid2895, signal peptidase complex (18kD), signal peptidase complex catalytic subunit SEC11A, SPase 18 kDa subunit, SPC18SEC11-like 1 (S. cerevisiae), SPCS4A1810012E07Rik | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 37-145 of human SEC11A (NP_055115.1). KGLMVITGSESPIVVVLSGSMEPAFHRGDLLFLTNRVEDPIRVGEIVVFRIEGREIPIVHRVLKIHEKQNGHIKFLTKGDNNAVDDRGLYKQGQHWLEKKDVVGRARGF | |
| 0.02 mL | |
| Cell Biology | |
| 23478 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction