missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPC24 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32105C
This item is not returnable.
View return policy
Description
SPC24 Polyclonal antibody specifically detects SPC24 in Human samples. It is validated for Western Blot
Specifications
| SPC24 | |
| Polyclonal | |
| Western Blot | |
| FLJ90806, hSpc24, kinetochore protein Spc24, SPBC24, SPC24, NDC80 kinetochore complex component, homolog (S. cerevisiae), spindle pole body component 24 homolog, spindle pole body component 24 homolog (S. cerevisiae) | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Spindle Pole Body Component 24 using the numbering given in entry NP_872319.1 (GeneID 147841). | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Cell Cycle and Replication | |
| 147841 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction