missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UPP2 Monoclonal Antibody (OTI9G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812319
This item is not returnable.
View return policy
Description
Catalyzes the reversible phosphorylytic cleavage of uridine and deoxyuridine to uracil and ribose- or deoxyribose-1-phosphate. The produced molecules are then utilized as carbon and energy sources or in the rescue of pyrimidine bases for nucleotide synthesis. Shows substrate specificity and accept uridine, deoxyuridine, and thymidine as well as the two pyrimidine nucleoside analogs 5-fluorouridine and 5-fluoro-2(′)-deoxyuridine as substrates.Specifications
| UPP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95045 | |
| UPP2 | |
| Full length human recombit protein of human UPP2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI9G9 | |
| Unconjugated | |
| UPP2 | |
| UDRPASE2, UP2, UPASE2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 151531 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction