missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPCS2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18429CL1
This item is not returnable.
View return policy
Description
SPCS2 Polyclonal antibody specifically detects SPCS2 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| SPCS2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9789 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| MGC117366, signal peptidase complex subunit 2, signal peptidase complex subunit 2 homolog (S. cerevisiae), SPC25 | |
| Recognizes region between residues Between Between 176 and 226.. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction