Learn More
SPCS2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-18429JF646
Code produit. 18546619
  • € 658.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
SPCS2 Polyclonal antibody specifically detects SPCS2 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Spécification
Spécification
SPCS2
Polyclonal
Western Blot
MGC117366, signal peptidase complex subunit 2, signal peptidase complex subunit 2 homolog (S. cerevisiae), SPC25
Recognizes region between residues Between Between 176 and 226..
0.1 mL
Primary
Human, Mouse
Purified
Western Blot
Janelia Fluor 646
50mM Sodium Borate
Rabbit
Affinity purified
RUO
9789
Store at 4°C in the dark.