missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPDYC Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98160AF488
This item is not returnable.
View return policy
Description
SPDYC Polyclonal antibody specifically detects SPDYC in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SPDYC | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 387778 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| hSpy/Ringo C, Rapid inducer of G2/M progression in oocytes C, RINGO C, Ringo2, speedy C, speedy homolog C (Xenopus laevis), speedy protein C | |
| Produced in rabbits immunized with E. coli-derived Human SPDYC fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction