missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPDYE4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555242
This item is not returnable.
View return policy
Description
Immunogen sequence: KCLGEIMASG QARPPFEEES PQPSTTVRSP EVVVDDEVPG PSAPWIDPSP QPQSLGLKRK SEWSD Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 38%.
Specifications
| SPDYE4 | |
| Polyclonal | |
| Unconjugated | |
| SPDYE4 | |
| putative Speedy protein E4; SPDYE4; speedy; speedy homolog E4; speedy protein E4; speedy/RINGO cell cycle regulator family member E4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 388333 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6NLX3 | |
| SPDYE4 | |
| Recombinant protein corresponding to Human SPDYE4. Recombinant protein control fragment (Product #RP-93502). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction